Merck
  • Influence of yeast mannoproteins in the aroma improvement of white wines.
Influence of yeast mannoproteins in the aroma improvement of white wines.
Journal of food science (2012-08-07)
M Juega, Y P Nunez, A V Carrascosa, A J Martinez-Rodriguez
ABSTRACT
In the present work, 3 different yeast strains (1, 2, and 3) were used to elaborate white wines using Albariño must. The concentration of polymeric mannose was determined using a method based on the mannoprotein precipitation, hydrolysis and analysis of sylylated mannose derivatives by gas chromatography. Wines elaborated with the strain 1 (W1) presented a higher mannoprotein concentration than the other wines. The analysis of the volatile composition of wines showed significant differences (P < 0.05) among them, being W1 which presented the highest concentration of aroma compounds, mainly terpens and norisoprenoids. The sensorial analysis of wines also showed that W1 had the best quality. The results obtained from this work demonstrate that mannoproteins could be involved in the behavior observed. Some evidences were obtained using a model wine, where 2 major terpens in W1 were preferentially retained by the colloids rich in mannoproteins released by strain 1. White wines elaborated with yeast strains overproducing mannoproteins could have better quality than others. Mannoproteins could contribute to aroma enhancement of Albariño white wines.
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Supelco
Geraniol, analytical standard
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Supelco
Linalool, analytical standard
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%