- An unexpected promiscuous activity of 4-oxalocrotonate tautomerase: the cis-trans isomerisation of nitrostyrene.
An unexpected promiscuous activity of 4-oxalocrotonate tautomerase: the cis-trans isomerisation of nitrostyrene.
Chembiochem : a European journal of chemical biology (2012-08-02)
Ellen Zandvoort, Edzard M Geertsema, Bert-Jan Baas, Wim J Quax, Gerrit J Poelarends
PMID22851288
ABSTRACT
Serendipitous switch: While exploring cis-nitrostyrene as a potential electrophile in Michael-type addition reactions catalysed by the enzyme 4-oxalocrotonate tautomerase (4-OT), it was unexpectedly found that 4-OT catalyses the isomerisation of cis-nitrostyrene to trans-nitrostyrene (k(cat) /K(m) = 1.9×10(3) M(-1) s(-1) ).