Merck
Iron-complexed adsorptive membrane for As(V) species in water.
Journal of hazardous materials (2012-07-28)
Rakesh N Shinde, Sadananda Das, R Acharya, N S Rajurkar, Ashok K Pandey
ABSTRACT
Selective preconcentration of a target analyte in the solid phase is an effective route not only to enhance detection limit of the conventional analytical method but also for elimination of interfering matrix. An adsorptive membrane was developed for selective preconcentration and quantification of ultra-trace (ppb) amounts of As(V) present in a variety of aqueous samples. The precursor membrane was prepared by UV-initiator induced graft polymerization of sulphate and phosphate bearing monomers (1:1 mol proportion) in pores of the host microporous poly(propylene) membrane. Fe(3+) ions were loaded in the precursor membrane to make it selective for As(V) ions. The presence of phosphate functional groups prevent leaching of Fe(3+) ions from the membrane when it comes in contact with solution like seawater having high ionic strength. The optimized membrane was characterized in terms of its physical structure, chemical structure and experimental conditions affecting As(V) uptake in the membrane. The possibility of quantifying total preconcentration of As content was also explored by converting As(III) to As(V). To quantify As(V), the membrane samples were subjected to instrumental neutron activation analysis (INAA). The studies carried in the present work showed that quantification of inorganic arsenic species in natural water samples is easily possible in 2-3 ppb concentration range.
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt solution, 50 wt. % in H2O
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%