Merck
  • Capreomycin and hygromycin B modulate the catalytic activity of the delta ribozyme in a manner that depends on the protonation and complexation with Cu2+ ions of these antibiotics.
Capreomycin and hygromycin B modulate the catalytic activity of the delta ribozyme in a manner that depends on the protonation and complexation with Cu2+ ions of these antibiotics.
Dalton transactions (Cambridge, England : 2003) (2012-07-14)
Milena Szafraniec, Kamila Stokowa-Sołtys, Justyna Nagaj, Aleksandra Kasprowicz, Jan Wrzesiński, Małgorzata Jeżowska-Bojczuk, Jerzy Ciesiołka
ABSTRACT
Catalytic RNA molecules (ribozymes) have often been used for the testing of interactions of antibiotics with ribonucleic acids. We showed that the impact of capreomycin and hygromycin B on delta ribozyme catalysis might change dramatically, from stimulation to inhibition, depending on conditions. In order to evaluate possible mechanisms of modulation of the ribozyme catalytic activity we used our earlier data on species distribution for protonated forms of capreomycin and hygromycin B and their complexes with Cu(2+) ions at different pH values. We proposed that, upon inhibition, the protonated amino group of capreomycin was located in the ribozyme catalytic cleft interfering with binding catalytic Mg(2+). Such a mechanism was also supported by the results of ribozyme inhibition with capreomycin complexed with Cu(2+). The effects of stimulation of the delta ribozyme activity by capreomycin and hygromycin B were less pronounced than inhibition. Possibly, the amino functions of these antibiotics might be involved in a general acid-base catalysis performed by the ribozyme, acting as proton acceptors/donors.
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, lyophilized powder
Sigma-Aldrich
Hygromycin B solution from Streptomyces hygroscopicus, ≥60% (HPLC), 45-60 mg/mL in H2O, γ-irradiated
Sigma-Aldrich
Hygromycin B solution from Streptomyces hygroscopicus, ≥60% (HPLC), 45-60 mg/mL in H2O