Merck
  • Simple and reliable methods for the determination of three steroidal glycosides in the eight species of Solanum by reversed-phase HPLC coupled with diode array detection.
Simple and reliable methods for the determination of three steroidal glycosides in the eight species of Solanum by reversed-phase HPLC coupled with diode array detection.
Phytochemical analysis : PCA (2012-07-13)
Anupam Maurya, N Manika, Ram Kishore Verma, Subhash C Singh, Santosh Kumar Srivastava
ABSTRACT
Solanum species are important ingredients of many traditional Indian medicines and thus the quality control of their herbal formulations is of paramount concern. To establish a simple and effective high-performance liquid chromatographic (HPLC) method to evaluate the quality of Solanum species and their herbal formulations. A rapid, simple, sensitive, robust and reproducible HPLC method was developed for the determination of three steroidal glycosides (SG); indioside D, solamargine and α-solanine in eight species of the genus Solanum. The analytes were separated on a monolithic performance RP-18e column (100 mm × 4.6 mm i.d.) using a gradient elution of acetonitile-water containing 0.1% trifluoroacetic acid (TFA) as the mobile phase with a flow rate 0.4 mL/min and UV detection at λ 210 nm. The method was linear over the range 3-15 µg/mL (r > 9994). Accuracy, precision and repeatability were all within the required limits. The mean recoveries measured at the three concentrations were higher than 98.8% with RSD < 2% for the targets. The established method is simple and can be used as a tool for quality control of plant material or herbal formulation containing SG.
Sigma-Aldrich
α-Solanine, from potato sprouts, ≥95%