Merck
  • The affinity of histamine for Gq protein-coupled histamine H(1)-receptors is predominantly regulated by their internalization in human astrocytoma cells.
The affinity of histamine for Gq protein-coupled histamine H(1)-receptors is predominantly regulated by their internalization in human astrocytoma cells.
Journal of pharmacological sciences (2012-07-13)
Shigeru Hishinuma, Yuko Sato, Chizuru Akatsu, Masaru Shoji
ABSTRACT
We examined the regulatory mechanisms of the affinity of Gq protein-coupled histamine H(1)-receptors for histamine after histamine pretreatment in intact human U373 MG astrocytoma cells. In control cells, the displacement curves for histamine against the binding of 5 nM [(3)H]mepyramine, a radioligand for H(1)-receptors, showed the presence of two binding sites for histamine, that is, high and low affinity sites. Pretreatment with 0.1 mM histamine for 30 min at 37°C induced a significant reduction in the percentage of high affinity sites for histamine and a concomitant increase in the percentage of low affinity sites with no change in their pIC(50) values. These histamine-induced changes were insensitive to 30 µM KN-62, an inhibitor of Ca(2+)/calmodulin-dependent protein kinase II, but they were completely inhibited either by 0.4 mM ZnCl(2), an inhibitor of G protein-coupled receptor kinases (GRKs), or under hypertonic conditions, where clathrin-mediated endocytosis is known to be inhibited. These results suggest that histamine-induced conversion of high to low affinity sites for histamine is predominantly regulated by GRK/clathrin-mediated internalization of H(1)-receptors in human astrocytoma cells.
Sigma-Aldrich
KN-62, ≥95%, powder
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis