Merck
  • Rush-hour aromatic and chlorinated hydrocarbons in selected subway stations of Shanghai, China.
Rush-hour aromatic and chlorinated hydrocarbons in selected subway stations of Shanghai, China.
Journal of environmental sciences (China) (2012-07-13)
Yanli Zhang, Chunlei Li, Xinming Wang, Hai Guo, Yanli Feng, Jianmin Chen
ABSTRACT
Air samples were collected simultaneously at platform, mezzanine and outdoor in five typical stations of subway system in Shanghai, China using stainless steel canisters and analyzed by gas chromatography-mass selective detector (GC-MSD) after cryogenic preconcentration. Benzene, toluene, ethylbenzene and xylenes (BTEX) at the platforms and mezzanines inside the stations averaged (10.3 +/- 2.1), (38.7 +/- 9.0), (19.4 +/- 10.1) and (30.0 +/- 11.1) microg/m3, respectively; while trichloroethylene (TrCE), tetrachloroethylene (TeCE) and para-dichlorobenzene (pDCB), vinyl chloride and carbon tetrachloride were the most abundant chlorinated hydrocarbons inside the stations with average levels of (3.6 +/- 1.3), (1.3 +/- 0.5), (4.1 +/- 1.1), (2.2 +/- 1.1) and (1.2 +/- 0.3) microg/m3, respectively. Mean levels of major aromatic and chlorinated hydrocarbons were higher indoor (platforms and mezzanines) than outdoor with average indoor/outdoor (I/O) ratios of 1.1-9.5, whereas no significant indoor/outdoor differences were found except for benzene and TrCE. The highly significant mutual correlations (p < 0.01) for BTEX between indoor and outdoor and their significant correlation (p < 0.05) with methyl tert-butyl ether (MTBE), a marker of traffic-related emission without other indoor and outdoor sources, indicated that BTEX were introduced into the subway stations from indoor/outdoor air exchange and traffic emission should be their dominant source. TrCE and pDCB were mainly from indoor emission and TeCE might have both indoor emission sources and contribution from outdoor air, especially in the mezzanines.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Supelco
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Supelco
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Supelco
tert-Butyl methyl ether, analytical standard
Supelco
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%