Merck
Rapid and efficient access to secondary arylmethylamines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-07-07)
Nicolas Fleury-Brégeot, Jessica Raushel, Deidre L Sandrock, Spencer D Dreher, Gary A Molander
ABSTRACT
Ammoniomethyl trifluoroborates are very powerful reagents that can be used to access biologically relevant aryl- and heteroaryl-methylamine motifs via Suzuki-Miyaura cross-couplings. Until now, this method was limited to the production of tertiary and primary amines. The synthesis of a large array of secondary ammoniomethyltrifluoroborates has been achieved through a one step nucleophilic substitution reaction on the potassium bromomethyltrifluoroborate. Smooth cross-coupling conditions have been designed, based on the use of an aminobiphenyl palladium precatalyst, to couple these trifluoroborates efficiently with aryl bromides. This strategy offers a new way to access biologically relevant motifs and allows, with the previously developed methods, access to all three classes of aminomethylarenes.
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Methylamine hydrochloride, ≥98%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 2.0 M in THF