Merck
  • [Study on eliminating sodium nitrite and blocking nitrosamine synthesis by anthocyanin from skin of Alpinia galanga].
[Study on eliminating sodium nitrite and blocking nitrosamine synthesis by anthocyanin from skin of Alpinia galanga].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-06-29)
Junsheng Huang
ABSTRACT
This study was performed to determine the ability of eliminating sodium nitrite and blocking nitrosamine synthesis by anthocyanin from the skin of Alpinia galanga. purified by macroporous resin. The test was conducted under the condition of the simulated human gastric juice (pH 3.0, 37 degrees C) with VitC as positive control. The results showed that the max capability of eliminating sodium nitrite was 87.14%, which is 1.6 times sronger than that of VitC, and the max capability of blocking nitrosamine synthesis was 97.82%, which is 8 times sronger than that of VitC.
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%