Merck
  • A facile method for preparation of t-butyloxycarbonylamino acid p-nitroanilides.
A facile method for preparation of t-butyloxycarbonylamino acid p-nitroanilides.
International journal of peptide and protein research (1990-08-01)
K Noda, M Oda, M Sato, N Yoshida
ABSTRACT
A series of p-nitroanilides of t-butyloxycarbonylamino acids, including Boc-Trp, Boc-Asn, Boc-Gln, Boc-Ser(But), Boc-Thr(But), Boc-Asp(OBut), Boc-Lys(TFA), and Boc-His(Boc), were prepared conveniently by the mixed anhydride method using 2,2-dimethylpropanoic chloride (pivaloyl chloride). The products were obtained in 40-60% yields after purification by column chromatography on Sephadex LH-20. p-Nitroanilide of histidine was purified after deprotection of Boc-His(Boc)-pNA and obtained as H-His-pNA.2HCl.