Merck
  • Exposure to monocrotophos pesticide during sexual development causes the feminization/demasculinization of the reproductive traits and a reduction in the reproductive success of male guppies (Poecilia reticulata).
Exposure to monocrotophos pesticide during sexual development causes the feminization/demasculinization of the reproductive traits and a reduction in the reproductive success of male guppies (Poecilia reticulata).
Toxicology and applied pharmacology (2012-06-21)
Hua Tian, Yun Li, Wei Wang, Peng Wu, Shaoguo Ru
ABSTRACT
Monocrotophos is a highly toxic organophosphorus pesticide that has been confirmed to be an endocrine-disrupting chemical. To evaluate the influence of this pollutant on the reproductive system of male fish, we studied the sex steroid levels, reproductive traits, sex ratio, and reproductive success in male guppies (Poecilia reticulata) exposed to 40% monocrotophos pesticide at the nominal concentrations of 0.01, 0.10, and 1.00 mg/L for 90 days from birth to adulthood in a semi-static exposure system. Radioimmunoassay and western blot analyses demonstrated that the long-term exposure to monocrotophos pesticide during the sexual development of male guppies caused a significant increase in 17β-estradiol levels and consequently induced vitellogenin synthesis, suggesting the feminization of the males. Monocrotophos pesticide also caused a significant decrease in testosterone levels, which consequently inhibited testis growth and reduced the sperm count and the area and intensity of their sexually attractive orange spots, which collectively indicated the significant demasculinization of the male sexual characteristics. Furthermore, these changes in the sexual characteristics at the cellular and organ levels translated into ecologically important effects on the reproductive success at the individual level, as measured by a decrease in offspring production and survival rate. The present study provides the first evidence that monocrotophos pesticide can cause severe reproductive abnormalities in fish due to its endocrine-disrupting action.