Merck
  • Improvement of durability of an organic photocatalyst in p-xylene oxygenation by addition of a Cu(II) complex.
Improvement of durability of an organic photocatalyst in p-xylene oxygenation by addition of a Cu(II) complex.
Physical chemistry chemical physics : PCCP (2012-06-14)
Yusuke Yamada, Kazuki Maeda, Kei Ohkubo, Kenneth D Karlin, Shunichi Fukuzumi
ABSTRACT
The catalytic durability of an organic photocatalyst, 9-mesityl-10-methyl acridinium ion (Acr(+)-Mes), has been dramatically improved by the addition of [{tris(2-pyridylmethyl)amine}Cu(II)](ClO(4))(2) ([(tmpa)Cu(II)](2+)) in the photocatalytic oxygenation of p-xylene by molecular oxygen in acetonitrile. Such an improvement is not observed by the addition of Cu(ClO(4))(2) in the absence of organic ligands. The addition of [(tmpa)Cu](2+) in the reaction solution resulted in more than an 11 times higher turnover number (TON) compared with the TON obtained without [(tmpa)Cu(II)](2+). In the photocatalytic oxygenation, a stoichiometric amount of H(2)O(2) formation was observed in the absence of [(tmpa)Cu(II)](2+), however, much less H(2)O(2) formation was observed in the presence of [(tmpa)Cu(II)](2+). The photocatalytic mechanism was investigated by laser flash photolysis measurements in order to detect intermediates. The reaction of O(2)˙(-) with [(tmpa)Cu(II)](2+) monitored by UV-vis spectroscopy in propionitrile at 203 K suggested formation of [{(tmpa)Cu(II)}(2)O(2)](2+), a transformation which is crucial for the overall 4-electron reduction of molecular O(2) to water, and a key in the observed improvement in the catalytic durability of Acr(+)-Mes.
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
p-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%