Merck
Methods for the determination of plasma or tissue glutathione levels.
Methods in molecular biology (Clifton, N.J.) (2012-06-07)
Trent E Tipple, Lynette K Rogers
ABSTRACT
We present two different methods for determining levels of glutathione in complex biological samples and plasma. The DTNB/GR enzyme recycling method is sensitive and requires no specialized equipment. The HPLC method is particularly useful for situations in which sample amounts are limited. Detailed instructions for performing each method as well as the advantages and disadvantages of each are discussed in this chapter.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, ≥93% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes