Merck
  • An evaluation of a manganese bath system having a new geometry through MCNP modelling.
An evaluation of a manganese bath system having a new geometry through MCNP modelling.
Radiation protection dosimetry (2012-05-26)
Rahim Khabaz
ABSTRACT
In this study, an approximate symmetric cylindrical manganese bath system with equal diameter and height was appraised using a Monte Carlo simulation. For nine sizes of the tank filled with MnSO(4).H(2)O solution of three different concentrations, the necessary correction factors involved in the absolute measurement of neutron emission rate were determined by a detailed modelling of the MCNP4C code with the ENDF/B-VII.0 neutron cross section data library. The results obtained were also used to determine the optimum dimensions of the bath for each concentration of solution in the calibration of (241)Am-Be and (252)Cf sources. Also, the amount of gamma radiation produced as a result of (n,γ) the reaction with the nuclei of the manganese sulphate solution that escaped from the boundary of each tank was evaluated. This gamma can be important for the background in NaI(Tl) detectors and issues concerned with radiation protection.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) sulfate monohydrate, puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%