Merck
  • Silver triflate and palladium acetate co-catalyzed reaction of N'-(2-alkynylbenzylidene)hydrazide with N-allyl ynamide.
Silver triflate and palladium acetate co-catalyzed reaction of N'-(2-alkynylbenzylidene)hydrazide with N-allyl ynamide.
Organic letters (2012-05-24)
Ping Huang, Zhiyuan Chen, Qin Yang, Yiyuan Peng
ABSTRACT
A silver triflate and palladium acetate co-catalyzed reaction of N'-(2-alkynylbenzylidene)hydrazide with N-allyl ynamide is described, which generates 2-amino-H-pyrazolo[5,1-a]isoquinolines in good to excellent yield. The transformation proceeds with high efficiency through 6-endo cyclization, [3 + 2] cycloaddition, 3,3-sigmatropic rearrangement, and aromatization.
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)