Merck
  • Coumarin-trioxane hybrids: synthesis and evaluation as a new class of antimalarial scaffolds.
Coumarin-trioxane hybrids: synthesis and evaluation as a new class of antimalarial scaffolds.
Bioorganic & medicinal chemistry letters (2012-05-23)
Koneni V Sashidhara, Abdhesh Kumar, Ranga Prasad Dodda, Naikade Niraj Krishna, Pooja Agarwal, Kumkum Srivastava, S K Puri
ABSTRACT
First synthesis of novel coumarin-trioxane hybrids is reported. The synthesis was achieved via condensation of β-hydroxyhydroperoxides with coumarinic-aldehydes in presence of p-toluenesulfonic acid in good yields and the novel hybrids were evaluated for their antimalarial activity both in vitro and in vivo.
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium p-toluenesulfonate, 95%
Sigma-Aldrich
Silver p-toluenesulfonate, ≥99%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ReagentPlus®, ≥98%