Merck
  • High level production of 5-aminolevulinic acid by Propionibacterium acidipropionici grown in a low-cost medium.
High level production of 5-aminolevulinic acid by Propionibacterium acidipropionici grown in a low-cost medium.
Biotechnology letters (2012-05-12)
Rossukon Sonhom, Chockpisit Thepsithar, Boonsri Jongsareejit
ABSTRACT
5-Aminolevulinic acid (ALA) is an intermediate in the biosynthesis of tetrapyrroles. Its current production is expensive. We have developed a low-cost medium for Propionibacterium acidipropionici to produce extracellular ALA. When grown at 35 °C on a medium containing 3 % (w/v) food-grade sodium lactate supplemented with 18 g glycine/l, 4.05 g succinate/l, 1.8 g glucose/l, pH 7, it produced ALA up to 7.7 g/l over 6 days. Plant-growth promoting activity assays showed that the ALA was biologically active.
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)