Merck
  • An assessment of the ocular safety of excipient maleic acid following intravitreal injection in rabbits.
An assessment of the ocular safety of excipient maleic acid following intravitreal injection in rabbits.
Toxicologic pathology (2012-05-04)
Shirley A Aguirre, Walter Collette, Hovhannes J Gukasyan, Wenhu Huang
ABSTRACT
Maleic acid was formulated in 0.7% saline and injected intravitreally in rabbits in order to evaluate ocular safety and tolerability. Maleic acid was formulated within a narrow pH range (2-3), administered in a fixed volume (100 µl), and concentrations ranged from 0.00 to 2.00 mg/eye (0.00 to 12.30 mM vitreous). Ocular evaluations were conducted at 2, 4, and 8 days post injection. Ocular irritation responses were observed at doses from 0.50 mg/eye (3.07 mM vitreous) to 2.00 mg/eye (12.30 mM vitreous) and included conjunctival redness and scleral swelling. Chemosis was observed at 2.00 mg/eye (12.30 mM vitreous). Funduscopic evaluations revealed enlarged retinal blood vessels and optic disk swelling at doses ≥1.50 mg/eye (9.22 mM vitreous), retinal folds and retinal discoloration at 2.00 mg/eye (12.30 mM vitreous). Histopathologic evaluations on days 4 and 8 post injection revealed retinal degeneration at doses ≥1.0 mg/eye (6.15 mM vitreous), conjunctival inflammation at doses ≥1.5 mg/eye (9.22 mM vitreous), and retinal pigment epithelial hypertrophy, optic nerve demyelination, anterior chamber fluid, and conjunctival fibrosis at 2.00 mg/eye (12.30 mM vitreous) maleic acid. The data suggest that maleic acid formulations at ≥1.00 mg/eye (6.15 mM vitreous) were not suitable for intraocular indications.
Product Description
Supelco
Maleic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium maleate dibasic, purum p.a., anhydrous, ≥98.0% (NT)
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Maleic acid sodium salt, ~99%