Merck
  • Significant endothelin release in patients treated with foam sclerotherapy.
Significant endothelin release in patients treated with foam sclerotherapy.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.] (2012-05-01)
Alessandro Frullini, Maria Chiara Barsotti, Tatiana Santoni, Emiliano Duranti, Silvia Burchielli, Rossella Di Stefano
ABSTRACT
Foam sclerotherapy has been proven to be a safe and effective treatment for superficial venous insufficiency, but transient visual and neurologic disturbances continue to be reported. These side effects have been theorized to be related to the presence of air or gases in the sclerosing foam that results in "bubble" migration into the cerebral circulation. We present a differing hypothesis that significant amounts of endothelin are released from the treated veins, amounts capable of causing these complications. We tested the release of endothelin 1 (ET-1) in 12 rats after sclerotherapy with sodium tetradecyl sulfate (STS) in liquid and foam preparations. In 11 human subjects, we measured ET-1 in systemic circulation and in a draining vein after foam sclerotherapy with polidocanol. Rats treated with STS showed a significant increase in ET-1 levels 1 and 5 minutes after foam sclerotherapy. Patients treated with foam sclerotherapy showed a marked increase in ET-1 levels that correlated significantly with local ET-1 levels. Evidence of ET-1 release represents a plausible relationship explaining neurologic and visual disturbances reported after sclerotherapy.
Sigma-Aldrich
Brij® L23 solution, 30 % (w/v) in H2O
Sigma-Aldrich
Decaethylene glycol mono­dodecyl ether, nonionic surfactant
Sigma-Aldrich
Thesit®, for membrane research
Sigma-Aldrich
Tetradecyl sulfate sodium salt, 95%
Sigma-Aldrich
ECO Brij® L23
Sigma-Aldrich
ECO BRIJ® L4, average Mn ~362
Sigma-Aldrich
Brij® L23, suitable for Stein-Moore chromatography
Sigma-Aldrich
Brij® L4, average Mn ~362
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether