Merck
  • Differential pulse voltammetric determination of methyl parathion based on multiwalled carbon nanotubes-poly(acrylamide) nanocomposite film modified electrode.
Differential pulse voltammetric determination of methyl parathion based on multiwalled carbon nanotubes-poly(acrylamide) nanocomposite film modified electrode.
Journal of hazardous materials (2012-04-13)
Yanbo Zeng, Dajun Yu, Yanyan Yu, Tianshu Zhou, Guoyue Shi
ABSTRACT
A sensitive electrochemical differential pulse voltammetry method was developed for detecting methyl parathion based on multiwalled carbon nanotubes-poly(acrylamide) (MWCNTs-PAAM) nanocomposite film modified glassy carbon electrode. The novel MWCNTs-PAAM nanocomposite, containing high content of amide groups, was synthesized by PAAM polymerizing at the vinyl group functionalized MWCNTs surface using free radical polymerization. The MWCNTs-PAAM nanocomposite was characterized by Fourier transform infrared spectroscopy, thermal gravimetric analysis and scanning electron microscopy. Electrochemical behavior and interference studies of MWCNTs-PAAM/GCE for methyl parathion were investigated. The experimental results demonstrated that the MWCNTs-PAAM/GCE exhibited a high adsorption and strong affinity toward methyl parathion compared with some metal ions and nitroaromatic compounds, which exist in environmental samples. The adsorbed amount of methyl parathion on the MWCNTs-PAAM/GCE approached the equilibrium value upon 5 min adsorption time. A linear calibration curve for methyl parathion was obtained in the concentration range from 5.0×10(-9) to 1.0×10(-5) mol L(-1), with a detection limit of 2.0×10(-9) mol L(-1). The MWCNTs-PAAM/GCE was proved to be a suitable sensing tool for the fast, sensitive and selective determination of methyl parathion in environmental water samples.