Merck
  • Direct asymmetric Mannich reaction of phthalides: facile access to chiral substituted isoquinolines and isoquinolinones.
Direct asymmetric Mannich reaction of phthalides: facile access to chiral substituted isoquinolines and isoquinolinones.
Chemical communications (Cambridge, England) (2012-04-11)
Jie Luo, Haifei Wang, Fangrui Zhong, Jacek Kwiatkowski, Li-Wen Xu, Yixin Lu
ABSTRACT
The first Mannich reaction employing phthalides using a quinidine-based multifunctional catalyst has been developed. The reported method led to the synthesis of 3,3-disubstituted phthalide derivatives in excellent yields, with good diastereo- and enantioselectivities. Convenient synthesis of chiral isoquinolinones and isoquinolines has also been demonstrated.
Sigma-Aldrich
Phthalide, 98%