Merck
  • Porous SiO₂/MgF₂ broadband antireflection coatings for superstrate-type silicon-based tandem cells.
Porous SiO₂/MgF₂ broadband antireflection coatings for superstrate-type silicon-based tandem cells.
Optics express (2012-03-29)
Na-Fu Wang, Ting-Wei Kuo, Yu-Zen Tsai, Shi-Xiong Lin, Pin-Kun Hung, Chiung-Lin Lin, Mau-Phon Houng
ABSTRACT
The purpose of this study is to reduce the glass substrate reflectivity over a wide spectral range (400-1200 nm) without having high reflectivity in the near-infrared region. After making porous SiO₂/MgF₂ double-layer antireflection (DLAR) thin film structure, the superstrate-type silicon-based tandem cells are added. In comparison to having only silicon-based tandem solar cells, the short-circuit current density has improved by 6.82% when porous SiO₂/MgF₂ DLAR thin film is applied to silicon-based tandem solar cells. This study has demonstrated that porous SiO₂/MgF₂ DLAR thin film structure provides antireflection properties over a broad spectral range (400-1200 nm) without having high reflectivity at near-infrared wavelengths.
Sigma-Aldrich
Magnesium fluoride, random crystals, optical grade, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium fluoride, technical grade
Sigma-Aldrich
Magnesium fluoride, pieces, 3-6 mm, 99.9% trace metals basis (excluding Na)