Merck
CoBr2-TMTU-zinc catalysed-Pauson-Khand reaction.
Chemical communications (Cambridge, England) (2012-03-24)
Yuefan Wang, Lingmin Xu, Ruocheng Yu, Jiahua Chen, Zhen Yang
ABSTRACT
A cobalt-TMTU complex, derived from the in situ reduction of CoBr(2) with Zn in the presence of TMTU, can catalyze Pauson-Khand reaction at a balloon pressure of CO, which enables the synthesis of structurally diverse cyclopentenones. This catalytic system works efficiently for both intermolecular and intramolecular PK reactions.
Sigma-Aldrich
Tetramethylthiourea, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Cobalt(II) bromide, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Cobalt(II) bromide, 99%
Pricing and availability is not currently available.