Merck
  • Vitrification-based cryopreservation of shoot-tips of Pinus kesiya Royle ex. Gord.
Vitrification-based cryopreservation of shoot-tips of Pinus kesiya Royle ex. Gord.
Cryo letters (2012-03-22)
V Kalita, H Choudhury, S Kumaria, P Tandon
ABSTRACT
The present investigation was aimed at developing a protocol for long-term preservation of germplasm of Pinus kesiya Royle ex. Gord. through vitrification. Some of the critical components affecting explant tolerance to cryopreservation, such as effects of preculture, vitrification solutions, exposure time to vitrification solutions, volume of vitrification solution and its toxicity, washing of vitrified tissues after thawing, were analysed. The results showed that shoot regrowth of P. kesiya shoot-tips was considerably affected when exposed to cryoprotectants for longer periods of time (longer than 10 min). Among different vitrification solutions studied, maximum survival (76 percent) of shoot-tips was achieved with mVSL (using 0.6 ml of the solution) in MS basal medium containing 4.0 mg l-1 N6-benzyladenine (BA).
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Sigma-Aldrich
Kinetin, suitable for plant cell culture, BioReagent, amorphous powder
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Supelco
6-Benzylaminopurine, PESTANAL®, analytical standard
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture