Merck
  • A new process for nickel ammonium disulfate production from ash of the hyperaccumulating plant Alyssum murale.
A new process for nickel ammonium disulfate production from ash of the hyperaccumulating plant Alyssum murale.
The Science of the total environment (2012-03-13)
R Barbaroux, E Plasari, G Mercier, M O Simonnot, J L Morel, J F Blais
ABSTRACT
The extraction of nickel (Ni) from ultramafic soils by phytomining can be achieved using Alyssum murale cultures. This study presents a new process for the valorization of Ni accumulated by this plant through the production of a Ni ammonium disulfate salt (Ni(NH(4))(2)(SO(4))(2).6H(2)O). The process comprises an initial leaching of the ashes of A. murale with a sulphuric acid solution (1.9 M H(2)SO(4), T=95 °C, t=240 min, TS=150 g ash L(-1)), producing a leachate rich in Ni (10.2 g Ni L(-1); 96% Ni solubilisation), Mg, P, K, Fe, Ca and Al. The pH of the acid leachate is increased to 5.0 with NaOH (5M), followed by an evaporation step which produced a purified solution rich in Ni (21.3 g NiL(-1)) and an iron hydroxide precipitate. The cold crystallization (T=2 °C, t=6h) of this solution by the stoichiometric addition (× 1.2) of ammonium sulfate generates a Ni ammonium disulfate salt, containing 13.2% Ni, that is potentially valuable to industry.
Sigma-Aldrich
Ammonium nickel(II) sulfate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium nickel(II) sulfate hexahydrate, ≥98%