Merck
  • Catalytic enantioselective desymmetrization of meso-glutaric anhydrides using a stable Ni2-Schiff base catalyst.
Catalytic enantioselective desymmetrization of meso-glutaric anhydrides using a stable Ni2-Schiff base catalyst.
Organic letters (2012-02-22)
Purushothaman Gopinath, Takumi Watanabe, Masakatsu Shibasaki
ABSTRACT
We describe the desymmetrization of meso-glutaric anhydrides to chiral hemiesters using a bench-stable homodinuclear Ni(2)-(Schiff base) complex as the catalyst in good to excellent yield (up to 99%) and enantioselectivity (up to 94%). Using the opposite enantiomer of the catalyst, we obtained the same yield and enantioselectivity with the opposite configuration, thereby gaining access to both hemiester enantiomers.
Sigma-Aldrich
Glutaric anhydride, 95%