Merck
  • Optical characteristics of YVO4:Eu3+ phosphor in close proximity to Ag nanofilm: emitting layer for mirror-type displays.
Optical characteristics of YVO4:Eu3+ phosphor in close proximity to Ag nanofilm: emitting layer for mirror-type displays.
Optics express (2012-02-15)
Cheol Jang, Seong Min Lee, Kyung Cheol Choi
ABSTRACT
We demonstrate the optical characteristics of YVO4:Eu3+ phosphor in close proximity to Ag nanofilm to create a highly efficient emitting layer in mirror-type self-emissive displays. The propagating surface plasmon mode induced between the dielectric layer (MgO) and the Ag nanofilm activates the electric dipole transition of Eu3+ ions. The transmittance of a 100 nm-thick Ag nanofilm is zero in the visible wavelength range, making this nanofilm a good reflector in the visible wavelength range and capable of fulfilling a mirror function. The emission of an YVO4:Eu3+ phosphor layer with a 100 nm-thick Ag nanofilm was enhanced to the point that it was eight times higher than that of a reference sample without Ag nanofilm. Therefore, the present work shows potential for application to mirror-type displays with high luminous efficacy.
Sigma-Aldrich
Yttrium(III) oxide, dispersion, 10 wt. % in isopropanol, nanoparticles, <100 nm (DLS), ≥99.9% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, nanopowder, <50 nm particle size
Sigma-Aldrich
Yttrium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, 99.999% trace metals basis