Merck
  • Secondary healing after removal of large keratocystic odontogenic tumor in the mandible: enucleation followed by open packing of iodoform gauze.
Secondary healing after removal of large keratocystic odontogenic tumor in the mandible: enucleation followed by open packing of iodoform gauze.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2012-02-15)
Hongzhi Zhou, Rui Hou, Qin Ma, Kenneth Wu, Yuxiang Ding, Ruifeng Qin, Kaijin Hu
ABSTRACT
The aim of this retrospective study was to present the findings of an open packing method after enucleation of large keratocystic odontogenic tumors (KCOTs) in the mandible. We performed a retrospective case series study of 27 patients with KCOTs larger than 5 cm treated at our institution between September 2003 and September 2008. A conservative surgical treatment was applied, which involved enucleation of the primary lesion and open packing of the residual osseous defect with iodoform gauze for secondary healing. Bone regeneration, tumor recurrence, and surgical complications were observed and analyzed. We used the Ļ‡(2) test and Pearson correlation coefficient for statistical analysis. The postoperative follow-up time was 52.3 months on average (range, 24 to 84 months). The packing gauze was changed every 2 weeks after enucleation, and the total duration for packing was 10.2 months on average (range, 7-15 months). Bone regeneration and satisfactory secondary healing were observed clinically and radiographically after treatment. Only 1 case had a recurrence 6 months after initial treatment, which was attributed to insufficient bony unroofing during enucleation. The recurrent lesion was re-treated by the same method, and no recurrence occurred in the following 6 years. No serious complications from this method of treatment were observed. No significant variables were found to be related to the recurrence. Enucleation with subsequent open packing was shown to be a conservative and comfortable treatment for patients and appears to be an effective choice for the management of large KCOTs in the mandible.
Sigma-Aldrich
Iodoform, 99%