Merck
  • Inorganic nitrate ingestion improves vascular compliance but does not alter flow-mediated dilatation in healthy volunteers.
Inorganic nitrate ingestion improves vascular compliance but does not alter flow-mediated dilatation in healthy volunteers.
Nitric oxide : biology and chemistry (2012-01-31)
M Bahra, V Kapil, V Pearl, S Ghosh, A Ahluwalia
ABSTRACT
Ingestion of inorganic nitrate elevates blood and tissue levels of nitrite via bioconversion in the entero-salivary circulation. Nitrite is converted to NO in the circulation, and it is this phenomenon that is thought to underlie the beneficial effects of inorganic nitrate in humans. Our previous studies have demonstrated that oral ingestion of inorganic nitrate decreases blood pressure and inhibits the transient endothelial dysfunction caused by ischaemia-reperfusion injury in healthy volunteers. However, whether inorganic nitrate might improve endothelial function per se in the absence of a pathogenic stimulus and whether this might contribute to the blood pressure lowering effects is yet unknown. We conducted a randomised, double-blind, crossover study in 14 healthy volunteers to determine the effects of oral inorganic nitrate (8 mmol KNO(3)) vs. placebo (8 mmol KCl) on endothelial function, measured by flow-mediated dilatation (FMD) of the brachial artery, prior to and 3h following capsule ingestion. In addition, blood pressure (BP) was measured and aortic pulse wave velocity (aPWV) determined. Finally, blood, saliva and urine samples were collected for chemiluminescence analysis of [nitrite] and [nitrate] prior to and 3h following interventions. Inorganic nitrate supplementation had no effect on endothelial function in healthy volunteers (6.9±1.1% pre- to 7.1±1.1% post-KNO(3)). Despite this, there was a significant elevation of plasma [nitrite] (0.4±0.1 μM pre- to 0.7±0.2 μM post-KNO(3), p<0.001). In addition these changes in [nitrite] were associated with a decrease in systolic BP (116.9±3.8mm Hg pre- vs. 112.1±3.4 mm Hg post-KNO(3), p<0.05) and aPWV (6.5±0.1 m/s pre- to 6.2±0.1 post-KNO(3), p<0.01). In contrast KCl capsules had no effect on any of the parameters measured. These findings demonstrate that although inorganic nitrate ingestion does not alter endothelial function per se, it does appear to improve blood flow, in combination with a reduction in blood pressure. It is likely that these changes are due to the intra-vascular production of NO.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%