Merck
  • Chiral recognition of dapoxetine enantiomers with methylated-gamma-cyclodextrin: a validated capillary electrophoresis method.
Chiral recognition of dapoxetine enantiomers with methylated-gamma-cyclodextrin: a validated capillary electrophoresis method.
Journal of pharmaceutical and biomedical analysis (2012-01-28)
Gábor Neumajer, Tamás Sohajda, András Darcsi, Gergő Tóth, Lajos Szente, Béla Noszál, Szabolcs Béni
ABSTRACT
The enantiomers of dapoxetine, a serotonin transporter inhibitor for the treatment of premature ejaculation have been separated by cyclodextrin modified capillary zone electrophoresis using uncoated fused-silica capillary. Over 20 cyclodextrins were screened as chiral selectors, investigating the stability of the inclusion complexes and enantioseparating properties. According to the preliminary experiments as chiral selector randomly methylated-γ-cyclodextrin was chosen. The basic chemical and instrumental parameters of enantioseparation as concentration of buffer, chiral selector and organic additive, pH, temperature and applied voltage were optimized afterwards using an orthogonal experimental design. Using this methodology not only the optimal parameter values for chiral separation (15 °C, +15 kV, 70 mM acetate, 20 v/v% MeOH, pH*=4.5, 3 mM methylated-γ-CyD) but also the significance order of factors on resolution was determined. Applying these parameters an optimal resolution of 7.01 was achieved. The optimized method was then validated according to the ICH guideline Q2 (R1) with regard to repeatability, linearity range, LOD, LOQ, accuracy and robustness.
Sigma-Aldrich
γ-Cyclodextrin, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥90.0% cyclodextrin basis (HPLC)