Merck
  • The efficacy of monepantel against naturally acquired inhibited and developing fourth-stage larvae of Teladorsagia circumcincta in sheep in the United Kingdom.
The efficacy of monepantel against naturally acquired inhibited and developing fourth-stage larvae of Teladorsagia circumcincta in sheep in the United Kingdom.
Veterinary parasitology (2011-12-20)
C Ramage, D J Bartley, F Jackson, R Cody, B C Hosking
ABSTRACT
The inhibition of Teladorsagia and other nematode genera at the early fourth-stage is a biological process that allows the parasites to survive in their host in a dormant state when prevailing conditions may otherwise kill them or prevent their progeny from surviving in the external environment. A study was conducted in Scotland to evaluate the efficacy of monepantel, an amino-acetonitrile derivative, against natural infections of inhibited fourth-stage Teladorsagia spp. larvae. At necropsy it was determined that the untreated control sheep were additionally infected with developing fourth-stage Teladorsagia spp. larvae and this is the first published evidence on the efficacy of monepantel against natural infections of this parasite and stage. The study sheep, which had grazed on naturally contaminated pastures since birth, were transferred to indoor housing after a subset of animals was examined to confirm the presence of inhibited larvae within the study population prior to the experiment. After 14 days of housing, monepantel was orally administered at 2.5 mg/kg to half of the animals. The sheep were necropsied seven days later and their parasite burdens recovered for the determination of efficacy, which was 99.7% for the inhibited stages and 99.3% for the developing fourth-stages. In conclusion, monepantel dosed orally at 2.5 mg/kg is a highly effective treatment against naturally acquired infections of inhibited and developing fourth-stage larvae of Teladorsagia spp.