Merck
  • Surface potential of methyl isobutyl carbinol adsorption layer at the air/water interface.
Surface potential of methyl isobutyl carbinol adsorption layer at the air/water interface.
The journal of physical chemistry. B (2011-12-17)
Chi M Phan, Hiromichi Nakahara, Osamu Shibata, Yoshikiyo Moroi, Thu N Le, Ha M Ang
ABSTRACT
The surface potential (ΔV) and surface tension (γ) of MIBC (methyl isobutyl carbinol) were measured on the subphase of pure water and electrolyte solutions (NaCl at 0.02 and 2 M). In contrast to ionic surfactants, it was found that surface potential gradually increased with MIBC concentration. The ΔV curves were strongly influenced by the presence of NaCl. The available model in literature, in which surface potential is linearly proportional to surface excess, failed to describe the experimental data. Consequently, a new model, employing a partial charge of alcohol adsorption layer, was proposed. The new model predicted the experimental data consistently for MIBC in different NaCl solutions. However, the model required additional information for ionic impurity to predict adsorption in the absence of electrolyte. Such inclusion of impurities is, however, unnecessary for industrial applications. The modeling results successfully quantify the influence of electrolytes on surface potential of MIBC, which is critical for froth stability.
Sigma-Aldrich
4-Methyl-2-pentanol, 98%