Merck
  • Effects of different sweet preloads on incretin hormone secretion, gastric emptying, and postprandial glycemia in healthy humans.
Effects of different sweet preloads on incretin hormone secretion, gastric emptying, and postprandial glycemia in healthy humans.
The American journal of clinical nutrition (2011-12-14)
Tongzhi Wu, Beiyi R Zhao, Michelle J Bound, Helen L Checklin, Max Bellon, Tanya J Little, Richard L Young, Karen L Jones, Michael Horowitz, Christopher K Rayner
ABSTRACT
Macronutrient "preloads" can stimulate glucagon-like peptide 1 (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP), slow gastric emptying, and reduce postprandial glycemic excursions. After sweet preloads, these effects may be signaled by sodium-glucose cotransporter-1 (SGLT1), sweet taste receptors, or both. We determined the effects of 4 sweet preloads on GIP and GLP-1 release, gastric emptying, and postprandial glycemia. Ten healthy subjects were studied on 4 separate occasions each. A preload drink containing 40 g glucose, 40 g tagatose/isomalt mixture (TIM), 40 g 3-O-methylglucose (3OMG; a nonmetabolized substrate of SGLT1), or 60 mg sucralose was consumed 15 min before a (13)C-octanoic acid-labeled mashed potato meal. Blood glucose, plasma total GLP-1 and GIP, serum insulin, and gastric emptying were determined. Both glucose and 3OMG stimulated GLP-1 and GIP release in advance of the meal (each P < 0.05), whereas TIM and sucralose did not. The overall postprandial GLP-1 response was greater after glucose, 3OMG, and TIM than after sucralose (P < 0.05), albeit later after TIM than the other preloads. The blood glucose and insulin responses in the first 30 min after the meal were greatest after glucose (each P < 0.05). Gastric emptying was slower after both 3OMG and TIM than after sucralose (each P < 0.05). In healthy humans, SGLT1 substrates stimulate GLP-1 and GIP and slow gastric emptying, regardless of whether they are metabolized, whereas the artificial sweetener sucralose does not. Poorly absorbed sweet tastants (TIM), which probably expose a greater length of gut to nutrients, result in delayed GLP-1 secretion but not in delayed GIP release. These observations have the potential to optimize the use of preloads for glycemic control. This trial was registered at www.actr.org.au as ACTRN12611000775910.
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Sucralose, analytical standard
Sigma-Aldrich
D-(−)-Tagatose, ≥98% (HPLC)
USP
Isomalt, United States Pharmacopeia (USP) Reference Standard
Isomalt, European Pharmacopoeia (EP) Reference Standard