- Development of the enantioselective addition of ethyl diazoacetate to aldehydes: asymmetric synthesis of 1,2-diols.
Development of the enantioselective addition of ethyl diazoacetate to aldehydes: asymmetric synthesis of 1,2-diols.
Journal of the American Chemical Society (2011-11-18)
Barry M Trost, Sushant Malhotra, Philipp Koschker, Pascal Ellerbrock
PMID22088096
ABSTRACT
A novel synthetic strategy toward the asymmetric synthesis of vicinal diols bearing a tertiary center is presented. The method encompasses the dinuclear Mg-catalyzed asymmetric addition of ethyl diazoacetate into several aldehydes, oxidation of the diazo functionality, and diastereoselective alkyl transfer of various organometallics into the resulting chiral β-hydroxy-α-ketoesters to afford a diverse range of 1,2-diols in high yield, diastereoselectivity, and chirality transfer.