Merck
Ruthenium-catalyzed meta sulfonation of 2-phenylpyridines.
Journal of the American Chemical Society (2011-11-04)
Ourida Saidi, Jameel Marafie, Araminta E W Ledger, Po Man Liu, Mary F Mahon, Gabriele Kociok-Köhn, Michael K Whittlesey, Christopher G Frost
ABSTRACT
A selective catalytic meta sulfonation of 2-phenylpyridines was found to occur in the presence of (arene)ruthenium(II) complexes upon reaction with sulfonyl chlorides. The 2-pyridyl group facilitates the formation of a stable Ru-C(aryl) σ bond that induces a strong para-directing effect. Electrophilic aromatic substitution proceeds with the sulfonyl chloride to furnish a sulfone at the position meta to the chelating group. This new catalytic process offers access to atypical regioselectivity for reactions involving chelation-assisted cyclometalation.
Sigma-Aldrich
2-Phenylpyridine, 98%
Sigma-Aldrich
Sulfuryl chloride solution, 1.0 M in methylene chloride