Merck
  • (-)-Secoisolariciresinol attenuates high-fat diet-induced obesity in C57BL/6 mice.
(-)-Secoisolariciresinol attenuates high-fat diet-induced obesity in C57BL/6 mice.
Food & function (2011-10-28)
Shiori Tominaga, Kosuke Nishi, Sogo Nishimoto, Koichi Akiyama, Satoshi Yamauchi, Takuya Sugahara
ABSTRACT
Flaxseed lignan, secoisolariciresinol has been reported to possess health benefits. We previously synthesized each stereoisomer of secoisolariciresinol and found that (-)-secoisolariciresinol reduces lipid accumulation and induces adiponectin production in 3T3-L1 adipocytes. Here we show the effects of (-)-secoisolariciresinol on high-fat diet-induced obesity in C57BL/6 male mice. Oral administration of (-)-secoisolariciresinol for 28 consecutive days significantly suppressed the gain of body weight. Increased serum adiponectin level and decreased gene expression of fatty acid synthase and sterol regulatory element-binding protein-1c in liver, which are related to fatty acid synthesis, were observed in the mice orally administered with (-)-secoisolariciresinol. In addition, subcutaneous injection of (-)-secoisolariciresinol also significantly suppressed the gain of body weight. Serum leptin levels were significantly increased by treating with (-)-secoisolariciresinol or (-)-enterolactone. Subcutaneous injection of (-)-secoisolariciresinol, (-)-enterolactone, or (-)-enterodiol promoted gene expression of acyl-CoA oxidase, carnitine palmitoyl transferase-1, and peroxisome proliferator-activated receptor Ī±, which are related to Ī²-oxidation. Overall results suggest that (-)-secoisolariciresinol exerts a suppressive effect on the gain of body weight of mice fed a high-fat diet by inducing gene expression of adiponectin, resulting in the altered expression of various genes related to the synthesis and Ī²-oxidation of fatty acids.
Sigma-Aldrich
Secoisolariciresinol, ≥95.0% (HPLC)