Merck
  • Amino acids and peptides. XXV. Preparation of fibronectin-related peptide poly(ethylene glycol) hybrids and their inhibitory effect on experimental metastasis.
Amino acids and peptides. XXV. Preparation of fibronectin-related peptide poly(ethylene glycol) hybrids and their inhibitory effect on experimental metastasis.
Chemical & pharmaceutical bulletin (1995-12-01)
K Kawasaki, M Namikawa, Y Yamashiro, Y Iwai, T Hama, Y Tsutsumi, S Yamamoto, S Nakagawa, T Mayumi
ABSTRACT
Hybrids of fibronectin-related peptides [Arg-Gly-Asp (RGD), Arg-Gly-Asp-Ser (RGDS)] and poly(ethylene glycol) (PEG) were prepared and their inhibitory effects on experimental metastasis in mice were examined. The inhibitory effect of RGD was markedly potentiated by hybrid formation with poly(ethylene glycol) #6000. As to inhibitory effect, RGD was more potent than RGDS and RGD PEG hybrids were superior to RGDS PEG hybrids. Hybrid formation with PEG #6000 was more effective than that with PEG #4000.
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 2,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 2,000