Merck
  • Integration of high accuracy N-terminus identification in peptide sequencing and comparative protein analysis via isothiocyanate-based isotope labeling reagent with ESI ion-trap TOF MS.
Integration of high accuracy N-terminus identification in peptide sequencing and comparative protein analysis via isothiocyanate-based isotope labeling reagent with ESI ion-trap TOF MS.
Journal of the American Society for Mass Spectrometry (2011-09-29)
Jiapeng Leng, Haoyang Wang, Li Zhang, Jing Zhang, Hang Wang, Tingting Cai, Jinting Yao, Yinlong Guo
ABSTRACT
A multifunctional isothiocyanate-based isotope labeling reagent, [d (0)]-/[d (6)]-4,6-dimethoxy pyrimidine-2-isothiocyanate (DMPITC), has been developed for accurate N-terminus identification in peptide sequencing and comparative protein analysis by ESI Ion-trap TOF mass spectrometry. In contrast with the conventional labeling reagent phenyl isothiocyanate (PITC), DMPITC showed more desirable properties such as rapid labeling, sensitivity enhancement, and facilitating peptide sequencing. More significantly, DMPITC-based labeling strategy possessed the capacity of higher reliable N-terminus identification owning to the high-yield b(1) ion combined with the isotope validation of 6 Da. Meanwhile, it also showed potential in differentiating isomeric residues of leucine and isoleucine at N-terminus on the basis of the relative abundance ratios between the fragment ions of their respective b(1) ions. The strategy not only allows accurate interpretation for peptide but also ensures rapid and sensitive comparative analysis for protein by direct MS analysis. Using trypsin-digested bovine serum albumin (BSA), both peptide N-terminus identification and quantitative analysis were accomplished with high accuracy, efficiency, and reproducibility. The application of DMPITC-based labeling strategy is expected to serve as a promising tool for proteome research.
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid