Merck
  • Pseudo-five-component reaction between 3-formylchromones, meldrum's acid, isocyanides and primary arylamines: diversity-oriented synthesis of novel chromone-containing peptidomimetics.
Pseudo-five-component reaction between 3-formylchromones, meldrum's acid, isocyanides and primary arylamines: diversity-oriented synthesis of novel chromone-containing peptidomimetics.
ACS combinatorial science (2011-09-17)
Mohammad Bagher Teimouri, Peyman Akbari-Moghaddam, Golara Golbaghi
ABSTRACT
An efficient and practical method has been developed for the diversity-oriented synthesis of chromone-containing tripeptides via pseudo-five-component reaction between 3-formylchromones, Meldrum's acid, isocyanides and primary aromatic amines for the generation of a wide range of structurally interesting and pharmacologically significant compounds at ambient temperature. It is worth mentioning that in the course of this reaction, five new bonds (two C-C bonds, two C-N bonds and one C═O bond) are formed. In the present reaction three amide bonds are newly formed.
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%