Merck
  • An energy efficient and facile synthesis of high molecular weight polyesters using ketenes.
An energy efficient and facile synthesis of high molecular weight polyesters using ketenes.
Chemical communications (Cambridge, England) (2011-09-01)
Martin Wolffs, Matthew J Kade, Craig J Hawker
ABSTRACT
A facile, ketene-based strategy for the synthesis of polyesters from stable Meldrum's acid monomers has been developed which overcomes many issues associated with traditional step-growth procedures. A significant increase in polymerization efficiency is observed with only 10 min reaction time at 220 °C being needed to obtain high molecular weight polymers.
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%