Merck
  • Experimental and theoretical study of hydrogen atom abstraction from n-butane by lanthanum oxide cluster anions.
Experimental and theoretical study of hydrogen atom abstraction from n-butane by lanthanum oxide cluster anions.
The journal of physical chemistry. A (2011-08-13)
Bo Xu, Yan-Xia Zhao, Xiao-Na Li, Xun-Lei Ding, Sheng-Gui He
ABSTRACT
Lanthanum oxide cluster anions are prepared by laser ablation and reacted with n-C(4)H(10) in a fast flow reactor. A time-of-flight mass spectrometer is used to detect the cluster distribution before and after the reactions. (La(2)O(3))(m=1-3)OH(-) and La(3)O(7)H(-) are observed as products, which suggests the occurrence of hydrogen atom abstraction reactions: (La(2)O(3))(m=1-3)O(-) + n-C(4)H(10) → (La(2)O(3))(m=1-3)OH(-) + C(4)H(9) and La(3)O(7)(-) + n-C(4)H(10) → La(3)O(7)H(-) + C(4)H(9). Density functional theory (DFT) calculations are performed to study the structures and bonding properties of La(2)O(4)(-), La(3)O(7)(-), and La(4)O(7)(-) clusters. The calculated results show that each of La(2)O(4)(-) and La(4)O(7)(-) contains one oxygen-centered radical (O(-•)) which is responsible for the high reactivity toward n-C(4)H(10). La(3)O(7)(-) contains one oxygen-centered radical (O(-•)) and one superoxide unit (O(2)(-•)), and the O(-•) is responsible for its high reactivity toward n-C(4)H(10). The O(-•) and O(2)(-•) can be considered to be generated by the adsorption of an O(2) molecule onto the singlet La(3)O(5)(-) with electron transfer from a terminally bonded oxygen ion (O(2-)) to the O(2). This may help us understand the mechanism of the formation of O(-•) and O(2)(-•) radicals in lanthanum oxide systems. The reaction mechanisms of La(2)O(4)(-) + n-C(4)H(10) and La(3)O(7)(-) + n-C(4)H(10) are also studied by the DFT calculations, and the calculated results are in good agreement with the experimental observations.
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis