Merck
Bismuth iodoform paraffin paste: a review.
The Journal of laryngology and otology (2011-07-13)
G J Crossland, A P Bath
ABSTRACT
This article reviews the literature pertaining to bismuth iodoform paraffin paste. Bismuth iodoform paraffin paste is used in most otolaryngology departments on a daily basis. Questions about its properties are common in postgraduate otolaryngology examinations. This article reviews bismuth iodoform paraffin paste's current and historical usage, constituents, properties, side effects, and radiographic properties, and its alternatives in otological and rhinological practice.
Sigma-Aldrich
Bismuth subnitrate, meets USP testing specifications
Sigma-Aldrich
Bismuth(III) subnitrate, 98%
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Iodoform, 99%
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Bismuth, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bismuth, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.97%
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bismuth, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.97%
Bismuth, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.97%
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Bismuth, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%