Merck
  • Treatment of alcohols with tosyl chloride does not always lead to the formation of tosylates.
Treatment of alcohols with tosyl chloride does not always lead to the formation of tosylates.
Molecules (Basel, Switzerland) (2011-07-05)
Rui Ding, Yong He, Xiao Wang, Jingli Xu, Yurong Chen, Man Feng, Chuanmin Qi
ABSTRACT
Treatment of substituted benzyl alcohols with tosyl chloride resulted in the formation of the corresponding chlorides, not the usual tosylates. A series of experiments demonstrated that it was possible to predict whether chlorination or tosylation would occur for substituted benzyl alcohols and pyridine methanols. Treatment of electron withdrawing group-substituted benzyl alcohols with tosyl chloride gave the corresponding chlorides in moderate yields under mild conditions, which provided a simple way to directly prepare chlorides from alcohols.
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%