Merck
  • Extending carbon chain length of 1-butanol pathway for 1-hexanol synthesis from glucose by engineered Escherichia coli.
Extending carbon chain length of 1-butanol pathway for 1-hexanol synthesis from glucose by engineered Escherichia coli.
Journal of the American Chemical Society (2011-06-29)
Yasumasa Dekishima, Ethan I Lan, Claire R Shen, Kwang Myung Cho, James C Liao
ABSTRACT
An Escherichia coli strain was engineered to synthesize 1-hexanol from glucose by extending the coenzyme A (CoA)-dependent 1-butanol synthesis reaction sequence catalyzed by exogenous enzymes. The C4-acyl-CoA intermediates were first synthesized via acetyl-CoA acetyltransferase (AtoB), 3-hydroxybutyryl-CoA dehydrogenase (Hbd), crotonase (Crt), and trans-enoyl-CoA reductase (Ter) from various organisms. The butyryl-CoA synthesized was further extended to hexanoyl-CoA via β-ketothiolase (BktB), Hbd, Crt, and Ter. Finally, hexanoyl-CoA was reduced to yield 1-hexanol by aldehyde/alcohol dehydrogenase (AdhE2). Enzyme activities for the C6 intermediates were confirmed by assays using HPLC and GC. 1-Hexanol was secreted to the fermentation medium under anaerobic conditions. Furthermore, co-expressing formate dehydrogenase (Fdh) from Candida boidinii increased the 1-hexanol titer. This demonstration of 1-hexanol production by extending the 1-butanol pathway provides the possibility to produce other medium chain length alcohols using the same strategy.
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Hexyl alcohol, natural, ≥98%, FCC, FG
Sigma-Aldrich
1-Hexanol, reagent grade, 98%
Supelco
1-Hexanol, analytical standard
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
Hexyl alcohol, FCC, FG