Merck
  • Effect of pendimethalin and quizalofop on N2-fixing bacteria in relation to availability of nitrogen in a Typic Haplustept soil of West Bengal, India.
Effect of pendimethalin and quizalofop on N2-fixing bacteria in relation to availability of nitrogen in a Typic Haplustept soil of West Bengal, India.
Environmental monitoring and assessment (2011-06-16)
Amal C Das, Hemanta Nayek, S Devi Nongthombam
ABSTRACT
An experiment was conducted under laboratory conditions to investigate the effect of two systemic herbicides viz., pendimethalin and quizalofop, at their recommended field rates (1.0 kg and 50 g active ingredient ha(- 1), respectively) on the growth and activities of non-symbiotic N(2)-fixing bacteria in relation to mineralization and availability of nitrogen in a Typic Haplustept soil. Both the herbicides, either singly or in a combination, stimulated the growth and activities of N(2)-fixing bacteria resulting in higher mineralization and availability of nitrogen in soil. The single application of quizalofop increased the proliferation of aerobic non-symbiotic N(2)-fixing bacteria to the highest extent while that of pendimethalin exerted maximum stimulation to their N(2)-fixing capacity in soil. Both the herbicides, either alone or in a combination, did not have any significant difference in the stimulation of total nitrogen content and availability of exchangeable NH(4)(+) while the solubility of NO(3)(-) was highly manifested when the herbicides were applied separately in soil.
Product Description
Supelco
Pendimethalin, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Pendimethalin solution, 100 μg/mL in methanol, PESTANAL®, analytical standard
Pricing and availability is not currently available.