Merck
  • Molecular hybridization of 4-azahexacyclo[5.4.1.0(2,6).0(3,10).0(5,9).0(8,11)]dodecane-3-ol with sigma (σ) receptor ligands modulates off-target activity and subtype selectivity.
Molecular hybridization of 4-azahexacyclo[5.4.1.0(2,6).0(3,10).0(5,9).0(8,11)]dodecane-3-ol with sigma (σ) receptor ligands modulates off-target activity and subtype selectivity.
Bioorganic & medicinal chemistry letters (2011-05-11)
Samuel D Banister, Iman A Moussa, William T Jorgensen, Sook Wern Chua, Michael Kassiou
ABSTRACT
A series of N-substituted 4-azahexacyclo[5.4.1.0(2,6).0(3,10).0(5,9).0(8,11)]dodecan-3-ols incorporating the respective arylalkyl subunits from several known sigma (σ) receptor ligands were synthesized and evaluated for their affinity against σ receptors and dopamine receptors. The hybrid trishomocubane-derived ligands (4-6) showed good selectivity for σ(1) and σ(2) receptors over multiple dopamine receptors. The molecular hybrid obtained from haloperidol and 4-azahexacyclo[5.4.1.0(2,6).0(3,10).0(5,9).0(8,11)]dodecan-3-ol (4, σ(1)K(i)=27 nM, σ(2)K(i)=55 nM) showed reduced affinity for D(1)-D(5) dopamine receptors when compared to haloperidol itself. The compound with the greatest σ(1) affinity in the series, benzamide 4 (σ(1)K(i)=7.6 nM, σ(2)K(i)=225 nM) showed a complete reversal of the subtype selectivity displayed by the highly σ(2) selective parent benzamide, RHM-2 (3, σ(1)K(i)=10412 nM, σ(2)K(i)=13.3 nM).
Product Description
Supelco
1-Dodecanol, Selectophore, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%
Pricing and availability is not currently available.