Merck
  • Honeybees Apis mellifera can detect the scent of Mycobacterium tuberculosis.
Honeybees Apis mellifera can detect the scent of Mycobacterium tuberculosis.
Tuberculosis (Edinburgh, Scotland) (2011-05-07)
David M Suckling, Rachael L Sagar
ABSTRACT
The proboscis extension reflex in honeybees was evaluated for detection of tuberculosis. Restrained bees were tested with methyl phenylacetate, methyl p-anisate, and methyl nicotinate, previously identified from Mycobacterium tuberculosis cultures, to determine honeybee capacity for signature volatile detection. Methyl p-anisate and methyl phenylacetate were detectable over eight orders of magnitude, and honeybees showed proboscis extension response down to 0.1 pg loading of methyl p-anisate on filter paper. Potential exists for trained honeybees in non-invasive diagnostic tests for TB.
Sigma-Aldrich
Methyl phenylacetate, ReagentPlus®, ≥99%
Sigma-Aldrich
Methyl nicotinate, ≥99%, FG
Sigma-Aldrich
Methyl phenylacetate, ≥98%, FCC, FG
Sigma-Aldrich
Methyl nicotinate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Methyl nicotinate, 99%