Merck
  • Fast liquid chromatography-tandem mass spectrometry for the analysis of bisphenol A-diglycidyl ether, bisphenol F-diglycidyl ether and their derivatives in canned food and beverages.
Fast liquid chromatography-tandem mass spectrometry for the analysis of bisphenol A-diglycidyl ether, bisphenol F-diglycidyl ether and their derivatives in canned food and beverages.
Journal of chromatography. A (2011-02-22)
H Gallart-Ayala, E Moyano, M T Galceran
ABSTRACT
In this work a fast liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) method using a C18 Fused Core™ column, was developed for the simultaneous analysis of bisphenol A diglycidyl ether (BADGE), bisphenol A (2,3-dihydroxypropyl) glycidyl ether (BADGE·H(2)O), bisphenol A bis(2,3-dihydroxypropyl) ether (BADGE·2H(2)O), bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether (BADGE·HCl), bisphenol A bis(3-chloro-2-hydroxypropyl) ether (BADGE·2HCl) and bisphenol A (3-chloro-2-hydroxypropyl)(2,3-dihydroxypropyl ether) (BADGE·HCl·H(2)O) and bisphenol F diglycidyl ether (BFDGE), bisphenol F bis(2,3-dihydroxypropyl) ether (BFDGE·2H(2)O), bisphenol F bis(3-chloro-2-hydroxypropyl) ether (BFDGE·2HCl). The LC method was coupled with a triple quadrupole mass spectrometer, using an ESI source in positive mode and using the [M+NH(4)](+) adduct as precursor ion for tandem mass spectrometry experiments. The method developed was applied to the determination of these compounds in canned soft drinks and canned food. OASIS HLB solid phase extraction (SPE) cartridges were used for the analysis of soft drinks, while solid canned food was extracted with ethyl acetate. Method limits of quantitation ranged from 0.13 μgL(-1) to 1.6 μgL(-1) in soft drinks and 1.0 μgkg(-1) to 4.0 μgkg(-1) in food samples. BADGE·2H(2)O was detected in all the analyzed samples, while other BADGEs such as BADGE·H(2)O, BADGE·HCl·H(2)O, BADGE·HCl and BADGE·2HCl were also detected in canned foods.
Sigma-Aldrich
Bisphenol A diglycidyl ether
Sigma-Aldrich
Acetonitrile, for UHPLC, suitable for mass spectrometry (MS)
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Supelco
Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, analytical standard
Supelco
Bisphenol F bis(2,3-dihydroxypropyl) ether, para-para isomer, analytical standard