Merck
  • Effects of aging and mixed nonaqueous-phase liquid sources in soil systems on earthworm bioaccumulation, microbial degradation, sequestration, and aqueous desorption of pyrene.
Effects of aging and mixed nonaqueous-phase liquid sources in soil systems on earthworm bioaccumulation, microbial degradation, sequestration, and aqueous desorption of pyrene.
Environmental toxicology and chemistry (2011-02-11)
Elijah J Petersen, Jixin Tang, Walter J Weber
ABSTRACT
The effects of loading and aging pyrene in soils in the presence of four environmentally common nonaqueous-phase liquids (NAPLs) (hexadecane, 2,2,4,4,6,8,8-heptamethylnonane [HMN], toluene, and dimethyl phthalate [DMP]) on its subsequent desorption from those soils, earthworm accumulation, biodegradation, and extractability were tested by using two dissimilar soils. The presence of each of the four NAPLs increased fractions and rates of pyrene desorption, and hexadecane slowed the effects of aging on these same parameters. Loading with hexadecane and HMN caused earthworm accumulation of pyrene to decrease. These results contrast with generally observed faster desorption rates resulting from NAPL addition, suggesting that additional factors (e.g., association of pyrene with NAPL phases and NAPL toxicities to earthworms) may impact bioaccumulation. The presence of HMN and toluene increased pyrene biodegradation, whereas hexadecane and DMP had the opposite effects. These results correlate with changes in the extractability of pyrene from the soils. After aging and biodegradation, hexadecane and DMP substantially increased pyrene residues extractable by methanol and decreased nonextractable fractions, whereas HMN and toluene had the opposite effects.
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
2,2,4,4,6,8,8-Heptamethylnonane, 98%