Merck
  • Different patterns of neuronal activities in the infralimbic and prelimbic cortices and behavioral expression in response to two affective odors, 2,5-dihydro-2,4,5-trimethylthiazoline and a mixture of cis-3-hexenol and trans-2-hexenal, in the freely moving rat.
Different patterns of neuronal activities in the infralimbic and prelimbic cortices and behavioral expression in response to two affective odors, 2,5-dihydro-2,4,5-trimethylthiazoline and a mixture of cis-3-hexenol and trans-2-hexenal, in the freely moving rat.
Behavioural brain research (2010-12-15)
Y Nikaido, T Nakashima
ABSTRACT
The medial prefrontal cortex (mPFC) is involved in stimulus perception, attentional control, emotional behavior, and the stress response. These functions are thought to be mediated by the infralimbic (IL) and prelimbic (PL) subregions of mPFC; however, few studies have examined the roles of IL and PL cortices in olfactory cognition. In the present study, we investigated the acute effects of two odors, 2,5-dihydro-2,4,5-trimethylthiazoline (TMT) and a mixture of cis-3-hexenol and trans-2-hexenal (green odor: GO), on behavioral responses and IL and PL neuronal activities using extracellular single-unit recordings in a freely moving rat. We found that the total number of spike firings in IL and PL neurons did not change with 10s presentation of odors. TMT presentation induced significant changes in burst firing activity in IL and PL neurons, while GO presentation induced changes in burst firing only in IL neurons. In the temporal profile of the firing activity of IL neurons, TMT exposure induced transient activation and GO exposure induced sustained activation. Those of PL neurons showed sustained activation during TMT exposure and transient activations during GO exposure. GO exposure induced a stretch-attend posture, whereas TMT exposure induced immobility. Furthermore, multiple regression analysis indicated that the property of the odor and neuronal activities of IL and PL regions were correlated with behavioral responses. These findings reveal that olfaction-related neurons exist in IL and PL regions, and that the neurons in these regions might temporarily encode odor information in order to modulate motor outputs by tuning firing properties in the early stage of cognition according to the odor property.
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%
Supelco
cis-3-Hexen-1-ol, analytical standard
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
trans-2-Hexen-1-al, ≥95%, FCC, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
trans-2-Hexen-1-al, 98%